Anti-RAC1+RAC2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-6154R-A488
: Bioss
- Antibody type:Primary
- Antigen name:Rho Family small GTP binding Protein Rac1 and Rac2
- Antigen symbol:RAC1+RAC2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5879, 5880
- Antigen synonyms:Ras related C3 botulinum toxin substrate 2|Rac-1|RAC1_HUMAN|Ras related C3 botulinum toxin substrate 1|p21 Rac2|MIG5|p21 Rac1|Ras like protein TC25|PAK1|Rac 1|RAC2_HUMAN.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:115-145/192
- Purification:Purified by Protein A
- Pk:100 µl
The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Mar 2009].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RAC1+RAC2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat