Norway
Order Entry
Anti-PRUNE Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-6299R-CY7
 :  Bioss
Anti-PRUNE Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-6299R-CY7
 :  Bioss
 
kr 10 550,00
   
  1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    Prune Exopolyphosphatase
  • Antigen symbol:
    PRUNE
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    58497
  • Antigen synonyms:
    HTCD37|DRES-17|DRES17
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

PRUNE, the human homologue of the Drosophila gene, is located in 1q21.3, a region highly amplified in human sarcomas, malignant tumours of mesenchymal origin. Human prune (h-prune), a phosphoesterase DHH family appertaining protein, physically interacts with nm23-H1, a metastasis suppressor gene. h-prune is involved in cellular motility and metastasis formation. Metastatic breast cancers were found to overexpress h-prune; h-prune was also found to be highly expressed in colorectal and pancreatic cancers. Hence h-prune is considered useful as a marker for tumor aggressiveness.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRUNE
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat