Insoles, X´Max
Leverandør: Honeywell Safety Products
X´Max
DALL6000350/46EA 205 NOK
DALL6000350/46
Insoles, X´Max
Skoinnleggssåler
Insoles designed for the Bac'Run® shoe range.
  • Shock absorber heel
  • Absorbs perspiration, facilitates air circulation
  • Impregnated with an antibacterial agent
  • Anti-static properties