Multi-channel pipettes, mechanical, variable volume, PIPETMAN® L
Leverandør: Gilson
PIPETMAN® L
GILSFA10040CEA 18275 NOK
GILSFA10040C GILSFA10016C GILSFA10038C GILSFA10036C GILSFA10014C 613-5819 GILSFA10012C GILSFA10010C 613-6917 613-6916 GILSFA10013CG GILSFA10036CG GILSFA10037CG GILSFA10012CG GILSFA10011CG GILSFA10038CG GILSFA10010CG GILSFA10039CG 613-5821 GILSFA10016CG 613-5820 GILSFA10015CG GILSFA10035CG GILSFA10014CG GILSFA10040CG GILSFA10039C GILSFA10037C GILSFA10015C GILSFA10013C GILSFA10035C GILSFA10011C 613-5814 GILSFA10009CG 613-5818 GILSFA10009C 613-5817 613-5816 613-5815
Multi-channel pipettes, mechanical, variable volume, PIPETMAN® L
Pipetter Pipetter manuelle og elektroniske
This ergonomic air displacement pipette is designed to provide all day pipetting comfort. The redesigned handle is lightweight and the finger hook provides added comfort.
  • Ergonomic features: Balanced weight, curved finger-hook, contoured handle
  • Adjustable tip ejector button can be used by both right- and left-handed users
  • Easy to remove tip ejector enables easy cleaning; entire lower part is fully autoclavable
  • Internal track system ensures consistent volume levels across all channels when aspirating and dispensing
  • Volume locking system: Volume cannot be accidentally changed
  • Colour coded push button that corresponds to match Gilson PIPETMAN tip packages
  • Features a 2-D identification code for easy traceability; readable by standard scanners
Tip ejector, tip holder and connecting nut can be sterilised in the laboratory under the following conditions: Moist heat/121 °C/20 minutes/1 bar.
Maximum permissible errors are guaranteed only when these pipettes are used with the recommended PIPETMAN DIAMOND tips.