Avantor® Apex ODS, HPLC Columns
: Avantor
Avantor® Apex and Avantor® Apex II HPLC column phases are recommended for routine analyses.
- Conventional 100 Å pore size spherical silica
- Narrow particle distribution
- Controlled surface area
- USP L1
- Catalog No:
- 4M10310
- FH1310-2/N
- 4M15310
- 4M25310
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- USP code:
- L1
- L1
- L1
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Apex™ columns
- Column FormatPhaseColumn hardwarePore sizeParticle SizeØ int.Length
- 4M10310AnalyticalODSStainless steel100 Å5 µm4,60 mm100 mm1 * 1 Itemskr 7 850,00
Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - 4M15310AnalyticalODSStainless steel100 Å5 µm4,60 mm150 mm1 * 1 Itemskr 12 075,00
Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - 4M25310AnalyticalODSStainless steel100 Å5 µm4,60 mm250 mm1 * 1 Itemskr 10 400,00
Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact
Guard cartridges, may require HPLC holders and accessories
- Column FormatPhaseColumn hardwarePore sizeParticle SizeØ int.Length
- FH1310-2/NGuard cartridgeODSStainless steel100 Å5 µm3,00 mm10 mm1 * 3 Itemskr 5 025,00
Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact
Guard holders and accessories
- SeqDescriptionPkPrice
- 11PEEK coupler (for use with HI-161 or HI-150)1POR