Dehydrated culture media, superbroth, Molecular Biology Certified™
Leverandør: MP Biomedicals
ICNA113010075EA 1675 NOK
ICNA113010075
Dehydrated culture media, superbroth, Molecular Biology Certified™
Superbroth is a rich medium with high levels of tryptone and yeast extract used for obtaining high yields of E. coli cells and lambda bacteriophages.
  • Available in pack pouches
  • Molecular biology certified
Due to the high levels of peptides, amino acids, vitamins and cofactors high yields of lambda phage and phagemids in E. coli are obtained.