Labsit Laboratory Chairs and Stool
Supplier: bimos
New Product
Certificates
About this item
Thanks to its innovative Flex functions, Labsit chairs are perfectly suited for laboratory work. Labsit offers outstanding comfort, laboratory ergonomics and intuitive operation.
- The backrest with its 3-D flex function follows the user’s movements
- Seat upholstery covered with PU foam or artificial leather, soft and comfortable, washable and antibacterial
- Resistant to all of the usual disinfectants and chemicals in accordance with ISO 2812
- Minimal joints thanks to 'Hygienic Design' for simple cleaning and disinfection
- Backrest in plastic and seat shell in a choice of five colours
- Integrated handle ensures maximum mobility and simple handling
- Seat height adjustment by gas spring
- Free from pollutants: no toxins or emissions
Flex function of the front edge of the seat: The front edge of the seat automatically follows the forward-angled working position of the body, which is typical for laboratory work. This ensures optimal blood circulation and effectively prevents pressure pain and dead legs. Labsit, therefore, provides exceptional comfort and optimal ergonomics.
Certifications: Suitable for cleanroom Category 3 in accordance with ISO 14644-1.
Suitable for use in biotechnology laboratories: Safety classes S1, S2, and S3.
GMP conformity.
GS certified.
Suitable for use in biotechnology laboratories: Safety classes S1, S2, and S3.
GMP conformity.
GS certified.
Accessories information: LabClip: The clever LabClip system means that every chair can be easily personalised and allocated to a user or an area. This creates structure in the workplace and makes a significant contribution to protecting the chair from contamination in the laboratory. It can also feature a company logo.
Standard laboratory chairs
Product Information
- Catalog #Supplier #HeightFoot styleFoot ringPkAvailabilityPrice
- Black artificial leather and plastic orange backrestBlack artificial leather and plastic orange backrestBIMO9123MG013279529123MG01327952450 - 650 mmCastors1Log in for availability1 * 1 Itemskr 3 300,00Supplier Article Number: 9123MG01327952Local Article Number: BIMO9123MG01327952Specifications:New Product
Accessories
Specifications
Environmentally PreferableNew Product - Black PU foam and white backrestBlack PU foam and white backrest132-127491232000340353450 - 650 mmCastors1Log in for availability1 * 1 Itemskr 3 000,00Supplier Article Number: 91232000340353Local Article Number: BIMO91232000340353Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and white backrestBlack artificial leather and white backrestBIMO9123MG013403529123MG01340352450 - 650 mmCastors1Log in for availability1 * 1 Itemskr 3 300,00Supplier Article Number: 9123MG01340352Local Article Number: BIMO9123MG01340352Specifications:New Product
Accessories
Specifications
Environmentally PreferableNew Product - Black artificial leather and white backrestBlack artificial leather and white backrest132-12699123MG01340353450 - 650 mmCastors1Log in for availability1 * 1 Itemskr 3 000,00Supplier Article Number: 9123MG01340353Local Article Number: BIMO9123MG01340353Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and green backrestBlack artificial leather and green backrest132-12719123MG01328053450 - 650 mmCastors1Log in for availability1 * 1 Itemskr 3 000,00Supplier Article Number: 9123MG01328053Local Article Number: BIMO9123MG01328053Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and anthracite backrestBlack artificial leather and anthracite backrest132-12689123MG01328553450 - 650 mmCastors1Log in for availability1 * 1 Itemskr 3 000,00Supplier Article Number: 9123MG01328553Local Article Number: BIMO9123MG01328553Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and blue backrestBlack artificial leather and blue backrest132-12679123MG01327753450 - 650 mmCastors1Log in for availability1 * 1 Itemskr 3 000,00Supplier Article Number: 9123MG01327753Local Article Number: BIMO9123MG01327753Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and orange backrestBlack artificial leather and orange backrest132-12709123MG01327953450 - 650 mmCastors1Log in for availability1 * 1 Itemskr 3 000,00Supplier Article Number: 9123MG01327953Local Article Number: BIMO9123MG01327953Specifications:
Accessories
Specifications
Environmentally Preferable - Supertec black, plastic orange backrestSupertec black, plastic orange backrestBIMO9123SP013279529123SP01327952450 - 650 mmCastors1Log in for availability1 * 1 Itemskr 3 875,00Supplier Article Number: 9123SP01327952Local Article Number: BIMO9123SP01327952Specifications:New Product
Accessories
Specifications
Environmentally PreferableNew Product - Black PU foam and anthracite backrestBlack PU foam and anthracite backrest132-127391232000328553450 - 650 mmCastors1Log in for availability1 * 1 Itemskr 3 000,00Supplier Article Number: 91232000328553Local Article Number: BIMO91232000328553Specifications:
Accessories
Specifications
Environmentally Preferable - Black PU foam and green backrestBlack PU foam and green backrest132-127691232000328053450 - 650 mmCastors1Log in for availability1 * 1 Itemskr 3 000,00Supplier Article Number: 91232000328053Local Article Number: BIMO91232000328053Specifications:
Accessories
Specifications
Environmentally Preferable - Black PU foam and orange backrestBlack PU foam and orange backrest132-127591232000327953450 - 650 mmCastors1Log in for availability1 * 1 Itemskr 3 000,00Supplier Article Number: 91232000327953Local Article Number: BIMO91232000327953Specifications:
Accessories
Specifications
Environmentally Preferable - Black PU foam and blue backrestBlack PU foam and blue backrest132-127291232000327753450 - 650 mmCastors1Log in for availability1 * 1 Itemskr 3 000,00Supplier Article Number: 91232000327753Local Article Number: BIMO91232000327753Specifications:
Accessories
Specifications
Environmentally Preferable
High laboratory chairs
Product Information
- Catalog #Supplier #HeightFoot styleFoot ringPkAvailabilityPrice
- Black integral foam, blue backrest, aluminium base polishedBlack integral foam, blue backrest, aluminium base polishedBIMO9125200032775291252000327752560 - 810 mmStop and Go castors and foot ring+1Log in for availability1 * 1 Itemskr 4 550,00Supplier Article Number: 91252000327752Local Article Number: BIMO91252000327752Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and anthracite backrestBlack artificial leather and anthracite backrest132-12789121MG01328553520 - 770 mmGlides+1Log in for availability1 * 1 Itemskr 3 550,00Supplier Article Number: 9121MG01328553Local Article Number: BIMO9121MG01328553Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and anthracite backrestBlack artificial leather and anthracite backrest132-12889125MG01328553560 - 810 mmCastors, Stop and Go+1Log in for availability1 * 1 Itemskr 3 750,00Supplier Article Number: 9125MG01328553Local Article Number: BIMO9125MG01328553Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and blue backrestBlack artificial leather and blue backrest132-12779121MG01327753520 - 770 mmGlides+1Log in for availability1 * 1 Itemskr 3 550,00Supplier Article Number: 9121MG01327753Local Article Number: BIMO9121MG01327753Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and blue backrestBlack artificial leather and blue backrestBIMO9121MG013279529121MG01327952520 - 770 mmGlides+1Log in for availability1 * 1 Itemskr 3 975,00Supplier Article Number: 9121MG01327952Local Article Number: BIMO9121MG01327952Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and blue backrestBlack artificial leather and blue backrest132-12879125MG01327753560 - 810 mmCastors, Stop and Go+1Log in for availability1 * 1 Itemskr 3 750,00Supplier Article Number: 9125MG01327753Local Article Number: BIMO9125MG01327753Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and green backrestBlack artificial leather and green backrest132-12819121MG01328053520 - 770 mmGlides+1Log in for availability1 * 1 Itemskr 3 550,00Supplier Article Number: 9121MG01328053Local Article Number: BIMO9121MG01328053Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and green backrestBlack artificial leather and green backrest132-12919125MG01328053560 - 810 mmCastors, Stop and Go+1Log in for availability1 * 1 Itemskr 3 750,00Supplier Article Number: 9125MG01328053Local Article Number: BIMO9125MG01328053Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and orange backrestBlack artificial leather and orange backrest132-12809121MG01327953520 - 770 mmGlides+1Log in for availability1 * 1 Itemskr 3 550,00Supplier Article Number: 9121MG01327953Local Article Number: BIMO9121MG01327953Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and orange backrestBlack artificial leather and orange backrest132-12909125MG01327953560 - 810 mmCastors, Stop and Go+1Log in for availability1 * 1 Itemskr 3 750,00Supplier Article Number: 9125MG01327953Local Article Number: BIMO9125MG01327953Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and white backrestBlack artificial leather and white backrest132-12799121MG01340353520 - 770 mmGlides+1Log in for availability1 * 1 Itemskr 3 550,00Supplier Article Number: 9121MG01340353Local Article Number: BIMO9121MG01340353Specifications:
Accessories
Specifications
Environmentally Preferable - Black artificial leather and white backrestBlack artificial leather and white backrest132-12899125MG01340353560 - 810 mmCastors, Stop and Go+1Log in for availability1 * 1 Itemskr 3 750,00Supplier Article Number: 9125MG01340353Local Article Number: BIMO9125MG01340353Specifications:
Accessories
Specifications
Environmentally Preferable - Black integral foam, green backrest, aluminium base polishedBlack integral foam, green backrest, aluminium base polishedBIMO9125200032805291252000328052560 - 810 mmStop and Go castors and foot ring+1Log in for availability1 * 1 ItemsSupplier Article Number: 91252000328052Local Article Number: BIMO91252000328052Specifications:
Accessories
Specifications
Environmentally Preferable - Black PU foam and anthracite backrestBlack PU foam and anthracite backrest132-128391212000328553520 - 770 mmGlides+1Log in for availability1 * 1 Itemskr 3 550,00Supplier Article Number: 91212000328553Local Article Number: BIMO91212000328553Specifications:
Accessories
Specifications
Environmentally Preferable - Black PU foam and anthracite backrestBlack PU foam and anthracite backrest132-129391252000328553560 - 810 mmCastors, Stop and Go+1Log in for availability1 * 1 Itemskr 3 750,00Supplier Article Number: 91252000328553Local Article Number: BIMO91252000328553Specifications:
Accessories
Specifications
Environmentally Preferable - Black PU foam and blue backrestBlack PU foam and blue backrest132-128291212000327753520 - 770 mmGlides+1Log in for availability1 * 1 Itemskr 3 550,00Supplier Article Number: 91212000327753Local Article Number: BIMO91212000327753Specifications:
Accessories
Specifications
Environmentally Preferable - Black PU foam and blue backrestBlack PU foam and blue backrest132-129291252000327753560 - 810 mmCastors, Stop and Go+1Log in for availability1 * 1 Itemskr 3 750,00Supplier Article Number: 91252000327753Local Article Number: BIMO91252000327753Specifications:
Accessories
Specifications
Environmentally Preferable - Black PU foam and green backrestBlack PU foam and green backrest132-128691212000328053520 - 770 mmGlides+1Log in for availability1 * 1 Itemskr 3 550,00Supplier Article Number: 91212000328053Local Article Number: BIMO91212000328053Specifications:
Accessories
Specifications
Environmentally Preferable - Black PU foam and green backrestBlack PU foam and green backrest132-129691252000328053560 - 810 mmCastors, Stop and Go+1Log in for availability1 * 1 Itemskr 3 750,00Supplier Article Number: 91252000328053Local Article Number: BIMO91252000328053Specifications:
Accessories
Specifications
Environmentally Preferable - Black PU foam and orange backrestBlack PU foam and orange backrest132-128591212000327953520 - 770 mmGlides+1Log in for availability1 * 1 Itemskr 3 550,00Supplier Article Number: 91212000327953Local Article Number: BIMO91212000327953Specifications:
Accessories
Specifications
Environmentally Preferable - Black PU foam and orange backrestBlack PU foam and orange backrest132-129591252000327953560 - 810 mmCastors, Stop and Go+1Log in for availability1 * 1 Itemskr 3 750,00Supplier Article Number: 91252000327953Local Article Number: BIMO91252000327953Specifications:
Accessories
Specifications
Environmentally Preferable - Black PU foam and white backrestBlack PU foam and white backrest132-128491212000340353520 - 770 mmGlides+1Log in for availability1 * 1 Itemskr 3 550,00Supplier Article Number: 91212000340353Local Article Number: BIMO91212000340353Specifications:
Accessories
Specifications
Environmentally Preferable - Black PU foam and white backrestBlack PU foam and white backrest132-129491252000340353560 - 810 mmCastors, Stop and Go+1Log in for availability1 * 1 Itemskr 3 750,00Supplier Article Number: 91252000340353Local Article Number: BIMO91252000340353Specifications:
Accessories
Specifications
Environmentally Preferable - Fabric duotec ESD black backrestFabric duotec ESD black backrestBIMO9125E-98019125E-9801560 - 810 mmStop and Go castors and foot ring+1Log in for availability1 * 1 Itemskr 5 800,00Supplier Article Number: 9125E-9801Local Article Number: BIMO9125E-9801Specifications:
Accessories
Specifications
Environmentally Preferable - Magic synthetic leather black, white backrest, aluminium base polishedMagic synthetic leather black, white backrest, aluminium base polishedBIMO9125MG013403529125MG01340352560 - 810 mmStop and Go castors and foot ring+1Log in for availability1 * 1 Itemskr 4 550,00Supplier Article Number: 9125MG01340352Local Article Number: BIMO9125MG01340352Specifications:
Accessories
Specifications
Environmentally Preferable - Magic synthetic leather, black backrestMagic synthetic leather, black backrestBIMO9125E-MG019125E-MG01560 - 810 mmStop and Go castors and foot ring+1Log in for availability1 * 1 Itemskr 5 800,00Supplier Article Number: 9125E-MG01Local Article Number: BIMO9125E-MG01Specifications:
Accessories
Specifications
Environmentally Preferable - Supertec black, anthracite backrest, aluminium base polishedSupertec black, anthracite backrest, aluminium base polishedBIMO9125SP013285529125SP01328552560 - 810 mmStop and Go castors and foot ring+1Log in for availability1 * 1 Itemskr 4 875,00Supplier Article Number: 9125SP01328552Local Article Number: BIMO9125SP01328552Specifications:
Accessories
Specifications
Environmentally Preferable - Supertec black, white backrestSupertec black, white backrestBIMO9125-SP01-34039125-SP01-3403560 - 810 mmStop and Go castors and foot ring+1Log in for availability1 * 1 Itemskr 4 625,00Supplier Article Number: 9125-SP01-3403Local Article Number: BIMO9125-SP01-3403Specifications:
Accessories
Specifications
Environmentally Preferable - Supertec ESD schwarz, black backrestSupertec ESD schwarz, black backrestBIMO9125E-CP019125E-CP01560 - 810 mmStop and Go castors and foot ring+1Log in for availability1 * 1 Itemskr 6 500,00Supplier Article Number: 9125E-CP01Local Article Number: BIMO9125E-CP01Specifications:
Accessories
Specifications
Environmentally Preferable - Supertec black, green backrestSupertec black, green backrestBIMO9125-SP01-32809125-SP01-3280560 - 810 mmStop and Go castors and foot ring+1Log in for availability1 * 1 Itemskr 4 625,00Supplier Article Number: 9125-SP01-3280Local Article Number: BIMO9125-SP01-3280Specifications:
Accessories
Specifications
Environmentally Preferable - Magic synthetic leather black, green backrest, aluminium base polishedMagic synthetic leather black, green backrest, aluminium base polishedBIMO9125MG013280529125MG01328052560 - 810 mmStop and Go castors and foot ring+1Log in for availability1 * 1 ItemsSupplier Article Number: 9125MG01328052Local Article Number: BIMO9125MG01328052Specifications:
Accessories
Specifications
Environmentally Preferable - Supertec black, blue backrestSupertec black, blue backrestBIMO9125-SP01-32779125-SP01-3277560 - 810 mmStop and Go castors and foot ring+1Log in for availability1 * 1 Itemskr 4 625,00Supplier Article Number: 9125-SP01-3277Local Article Number: BIMO9125-SP01-3277Specifications:
Accessories
Specifications
Environmentally Preferable
Laboratory stools
Product Information
- Catalog #Supplier #HeightFoot styleFoot ringPkAvailabilityPrice
- Integral foam Labsit laboratory stool with glides and footringIntegral foam Labsit laboratory stool with glides and footringBIMO9122-20009122-2000520 - 770 mmGlides1Log in for availability1 * 1 Itemskr 2 700,00Supplier Article Number: 9122-2000Local Article Number: BIMO9122-2000Specifications:
Accessories
Specifications
Environmentally Preferable - Integral foam black with polyamide base black backrestIntegral foam black with polyamide base black backrest132-129791272000532217450 - 650 mmCastors1Log in for availability1 * 1 Itemskr 2 100,00Supplier Article Number: 91272000532217Local Article Number: BIMO91272000532217Specifications:
Accessories
Specifications
Environmentally Preferable