Norway
Order Entry
Disposable particle filtering respirators, FFP3, 8833
Disposable particle filtering respirators, FFP3, 8833
Catalog # 111-0451
Supplier:  3M
undefined
Disposable particle filtering respirators, FFP3, 8833
Catalog # 111-0451
Supplier:  3M
Price
kr 2 100,00
Log in for availability
Unit of Measure 1 * 10 Items
Specifications
  • Type:
    8833
  • Size:
    U*
  • Pk:
    10
  • Class:
    FFP3
Specifications
About this item
The robust shell and cup design of the mask conforms well to most face shapes and sizes, and maintains its shape during use. The excellent fit is provided by the convex shape, the nose clip, and the twin-strap design.
  • Cool Flow™ valve reduces heat build-up to offer comfortable protection, particularly in hot and humid conditions
  • Soft inner face-seal ring provides secure face fit and increases wearer comfort
  • Reliable, effective protection against fine particulates
  • Comfortable, lightweight off-the-face design
  • Red colour coded straps denoting FFP3 protection level
Certifications: EN 149:2001 FFP3, Maximum use level: Assigned Protection Factor: 20×WEL (Nominal Protection Factor 50×WEL).
Caution: *One size fits all