Norway
Order Entry
Cleaning disinfectant, oxivir plus spray
Cleaning disinfectant, oxivir plus spray
  DIVY100829236
 :  Diversey
undefined
Cleaning disinfectant, oxivir plus spray
  DIVY100829236
 :  Diversey
  • Description:
    Oxivir plus, disinfectant trigger spray
  • Packed:
    Case of 6×750 ml spray bottles
  • Pk:
    6 Bottle
Oxivir plus spray is a concentrated washing and disinfectant prepared on active oxygen base AHP. Ideal for textiles and hard surfaces.
  • Hospital grade disinfectant
  • Based on proprietary hydrogen peroxide technology (AHP®)
  • Disinfects in 5 minutes at 1:40 dilution
  • Excellent surface safety
  • Kills MRSA ,VRE and Norovirus and Canine Parvovirus