General purpose gloves, FineDex® 098-10
Leverandør: Honeywell Safety Products
FineDex®
DALL2009810-07EA 86 NOK
DALL2009810-07 DALL2009810-08 DALL2009810-09 DALL2009810-10
General purpose gloves, FineDex® 098-10
Hansker
These PVC gloves with cotton flock interior provide outstanding flexibility and dexterity. Ideal for handlings in oily environments.
  • Cotton flocked interior provides the user with greater comfort
  • Diamond finish is tailored for grasping dry and damp objects
  • Low resistance to ketones, aromatic and halogen hydrocarbons
  • Cuff helps to protect a large part of the forearm
Typical applications include: General maintenance, cleaning, collective catering, standard work in contact with chemicals, maintenance.
Sertifiseringer: EN 388 (3XXX), EN 374; Cat. II