Connectors, tubing, Opta® SFT
Leverandør: Sartorius
Opta® SFT
SART640FS014M-DEA 14575 NOK
SART640FS014M-D
Connectors, tubing, Opta® SFT
Fittings og kontakter
The Opta® SFT range of sterile connectors allows fast and reliable sterile connection and sterile fluid transfer between two separate, pre-sterilised process components in biopharmaceutical manufacturing operations.
  • Easy, robust, repeatable operation
  • Claimed size equivalent to tubing ID size with no flow restriction
  • Safe and robust tubing connections
The OPTA® SFT-I connectors can be integrated into Sartorius Stedim Biotech Fluid Management Technologies assemblies and are also available as individual divise for end-user assembly with silicone tubing.