Avantor® Alltima, HPLC Columns, C18
: Avantor
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
- Base deactivated silica phases
- Stable bonding for long column lifetime
- Multiple selectivity options
- USP L1
Classic reversed phase retention and selectivity. High quality hydrophobic general purpose C18.
- Catalog No:
- 96080/N
- HICH88352
- 96681/N
- 96080SP/N
- HICH88051
- HICH81143
- HICH88053
- HICH43851
- HICH88056
- HICH81412
- HICH11543
- HICH81383
- HICH81382
- HICH81387
- AL-5C18-150DE
- 5174010/N
- 96680/N
- HICH88055
- HICH88057
- AL-5C18-300DF
- HICH81388
- AL-5C18-300DE
- AL-5C18-10CP2
- HICH50605
- HICH88063
- AL-10C18-300DF
- HICH81105
- HICH88888
- HICH88370
- HICH88307
- HICH88371
- AL-5C18-250DF
- HICH81140
- HICH81142
- HICH43853
- AL-3C18-75D
- HICHAL-3C18-100SP7
- HICH970203
- HICH88334
- HICH88335
- HICH88383
- AL-10C18-300DE
- HICH88889
- AL-5C18-150DF
- HICH88059
- HICH88353
- HICH88052
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Surface area:
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- Column FormatPhaseColumn hardwarePore sizeParticle SizeØ int.
- ColumnColumnHICH88352AnalyticalC18Stainless steel100 Å3 µm2,10 mm1 * 1 Itemskr 10 050,00: HICH88352
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH43853AnalyticalC18Stainless steel100 Å3 µm1,00 mm1 * 1 Itemskr 11 350,00: HICH43853
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH88353AnalyticalC18Stainless steel100 Å3 µm2,10 mm1 * 1 Itemskr 9 450,00: HICH88353
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH81143AnalyticalC18Stainless steel100 Å3 µm3,00 mm1 * 1 Itemskr 12 425,00: HICH81143
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH43851AnalyticalC18Stainless steel100 Å3 µm4,60 mm1 * 1 Itemskr 6 650,00: HICH43851
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH81412AnalyticalC18Stainless steel100 Å3 µm4,60 mm1 * 1 Itemskr 9 650,00: HICH81412
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - Column: HICHAL-3C18-75D
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - Column-W (Waters column port)Column-W (Waters column port)HICH81383AnalyticalC18Stainless steel100 Å3 µm4,60 mm1 * 1 Itemskr 10 525,00: HICH81383
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - ColumnColumnHICH81382AnalyticalC18Stainless steel100 Å3 µm4,60 mm1 * 1 Itemskr 10 650,00: HICH81382
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH81387AnalyticalC18Stainless steel100 Å3 µm4,60 mm1 * 1 Itemskr 12 650,00: HICH81387
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - Column-W (Waters column port)Column-W (Waters column port)HICH81388AnalyticalC18Stainless steel100 Å3 µm4,60 mm1 * 1 Itemskr 13 125,00: HICH81388
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - ColumnColumnHICH50605Semi-preparativeC18Stainless steel100 Å3 µm7,00 mm1 * 1 Itemskr 10 350,00: HICH50605
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - ColumnColumnHICHAL-3C18-100SP7Semi-preparativeC18Stainless steel100 Å3 µm7,00 mm1 * 1 Itemskr 12 300,00: HICHAL-3C18-100SP7
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - ColumnColumnHICH88888AnalyticalC18Stainless steel100 Å5 µm2,10 mm1 * 1 Itemskr 9 450,00: HICH88888
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH88370AnalyticalC18Stainless steel100 Å5 µm2,10 mm1 * 1 Itemskr 11 950,00: HICH88370
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH88371AnalyticalC18Stainless steel100 Å5 µm2,10 mm1 * 1 Itemskr 12 875,00: HICH88371
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH81140AnalyticalC18Stainless steel100 Å5 µm3,00 mm1 * 1 Itemskr 12 875,00: HICH81140
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH81142AnalyticalC18Stainless steel100 Å5 µm3,00 mm1 * 1 Itemskr 12 525,00: HICH81142
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH88383AnalyticalC18Stainless steel100 Å5 µm3,20 mm1 * 1 Itemskr 12 525,00: HICH88383
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnAL-5C18-150DEAnalyticalC18Stainless steel100 Å5 µm3,90 mm1 * 1 Itemskr 11 450,00: HICHAL-5C18-150DE
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnAL-5C18-300DEAnalyticalC18Stainless steel100 Å5 µm3,90 mm1 * 1 Itemskr 13 125,00: HICHAL-5C18-300DE
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnAL-5C18-150DFAnalyticalC18Stainless steel100 Å5 µm4,00 mm1 * 1 Itemskr 11 450,00: HICHAL-5C18-150DF
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnAL-5C18-300DFAnalyticalC18Stainless steel100 Å5 µm4,00 mm1 * 1 Itemskr 13 350,00: HICHAL-5C18-300DF
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH88889AnalyticalC18Stainless steel100 Å5 µm4,60 mm1 * 1 Itemskr 9 600,00: HICH88889
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH88059AnalyticalC18Stainless steel100 Å5 µm4,60 mm1 * 1 Itemskr 9 725,00: HICH88059
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH88052AnalyticalC18Stainless steel100 Å5 µm4,60 mm1 * 1 Itemskr 12 425,00: HICH88052
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH88051AnalyticalC18PEEK100 Å5 µm4,60 mm1 * 1 Itemskr 11 175,00: HICH88051
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Column-W (Waters column port)Column-W (Waters column port)HICH88053AnalyticalC18Stainless steel100 Å5 µm4,60 mm1 * 1 Itemskr 12 650,00: HICH88053
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - ColumnColumnAnalyticalC18Stainless steel100 Å5 µm4,60 mm1 * 1 Itemskr 12 650,00: HICH88056
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - Validation kit (3 columns from 3 different batches)Validation kit (3 columns from 3 different batches)HICH11543Method Validation KitC18Stainless steel100 Å5 µm4,60 mm1 * 3 Itemskr 40 375,00: HICH11543
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Column: HICHAL-5C18-250DF
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnHICH88055AnalyticalC18PEEK100 Å5 µm4,60 mm1 * 1 Itemskr 13 700,00: HICH88055
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Column-W (Waters column port)Column-W (Waters column port)HICH88057AnalyticalC18Stainless steel100 Å5 µm4,60 mm1 * 1 Itemskr 13 000,00: HICH88057
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - ColumnColumnHICH88063Semi-preparativeC18Stainless steel100 Å5 µm10,00 mm1 * 1 Itemskr 26 450,00: HICH88063
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - ColumnColumnHICH81105PreparativeC18Stainless steel100 Å5 µm22,00 mm1 * 1 Itemskr 75 250,00: HICH81105
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification - ColumnColumnAL-10C18-300DEAnalyticalC18Stainless steel100 Å10 µm3,90 mm1 * 1 Itemskr 12 075,00: HICHAL-10C18-300DE
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - ColumnColumnAL-10C18-300DFAnalyticalC18Stainless steel100 Å10 µm4,00 mm1 * 1 Itemskr 13 350,00: HICHAL-10C18-300DF
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - ColumnColumnHICH88307AnalyticalC18Stainless steel100 Å10 µm4,60 mm1 * 1 Itemskr 11 450,00: HICH88307
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - ColumnColumnHICH88334Semi-preparativeC18Stainless steel100 Å10 µm10,00 mm1 * 1 Itemskr 22 825,00: HICH88334
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - ColumnColumnHICH88335PreparativeC18Stainless steel100 Å10 µm22,00 mm1 * 1 Itemskr 92 675,00: HICH88335
Specifications
Environmentally PreferableCarbon load16%EndcappedYesSurface area340 m²/gUSP codeL1Environmental certification
Guard cartridges
- Column FormatPhaseColumn hardwarePore sizeParticle SizeØ int.
- All-guard cartridge for 3,0 – 4,6 mm ID columns (Use with holder 80101/N and coupler HI-081)All-guard cartridge for 3,0 – 4,6 mm ID columns (Use with holder 80101/N and coupler HI-081)96080/NGuard cartridgeC18Stainless steel5 µm1 * 3 Itemskr 3 800,00: HICH96080/N
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedSurface areaUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - All-guard cartridge for 2,1 mm ID columns (Use with holder 80101/N and coupler HI-081)All-guard cartridge for 2,1 mm ID columns (Use with holder 80101/N and coupler HI-081)96681/NGuard cartridgeC18Stainless steel3 µm1 * 3 Itemskr 3 775,00: HICH96681/N
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedSurface areaUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - All-guard cartridge for 3,0 – 4,6 mm ID columns (Use with holder 80101/N and coupler HI-081)All-guard cartridge for 3,0 – 4,6 mm ID columns (Use with holder 80101/N and coupler HI-081)96080SP/NGuard cartridgeC18Stainless steel5 µm1 * 10 Itemskr 11 175,00: HICH96080SP/N
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedSurface areaUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - All-guard cartridge for 2,1 mm ID columns (Use with holder 80101/N and coupler HI-081)All-guard cartridge for 2,1 mm ID columns (Use with holder 80101/N and coupler HI-081)5174010/NGuard cartridgeC18Stainless steel5 µm1 * 10 Itemskr 12 175,00: HICH5174010/N
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedSurface areaUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - All-guard cartridge for 2,1 mm ID columns (Use with holder 80101/N and coupler HI-081)All-guard cartridge for 2,1 mm ID columns (Use with holder 80101/N and coupler HI-081)96680/NGuard cartridgeC18Stainless steel5 µm1 * 3 Itemskr 3 775,00: HICH96680/N
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedSurface areaUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Guard Cartridge for 10 - 22 mm ID columns (Use with holder GCH10 and coupler HI-081)Guard Cartridge for 10 - 22 mm ID columns (Use with holder GCH10 and coupler HI-081)AL-5C18-10CP2Guard cartridgeC18Stainless steel5 µm1 * 2 Items: HICHAL-5C18-10CP2
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedSurface areaUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Prep‐Guard columnPrep‐Guard columnHICH970203Guard columnC18Stainless steel10 µm1 * 1 Itemskr 6 875,00: HICH970203
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedSurface areaUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact