Anti-ACSL5 Goat Polyclonal Antibody
PRSI45-197
- Antigen name:Acyl-Coa Synthetase Long-Chain Family Member 5
- Antigen symbol:ACSL5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:ACS5|fatty-acid-Coenzyme A ligase|long-chain 5|long-chain acyl-CoA synthetase 5|long-chain fatty acid coenzyme A ligase 5|ACSL5|FACL5|ACS2|HGNC:16526|acyl-CoA synthetase long-chain family member 5
- Concentration:500 ug/mL
- Immunogen:ACSL5 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of ACSL5.
- Size:100 µg
- Pk:100 µG
About this item
ACSL5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ACSL5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ACSL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human