Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Chromium standard solution, for ICP, in 5% HNO₃, VHG
:
LGC Standards PROMOCHEM
Chromium standard solution, for ICP, in 5% HNO₃, VHG
:
LGC Standards PROMOCHEM
Product Information
Element
Symbol
Concentration
Matrix
Pk
PROOVHG-PCRN-50
VHG-PCRN-50
Chromium
Cr
1000 mg/L
5% HNO₃
50 mL
1 * 50 mL
-
+
1 * 50 mL
-
+
PROOVHG-PCRN-100
VHG-PCRN-100
Chromium
Cr
1000 mg/L
5% HNO₃
100 mL
1 * 100 mL
1 * 100 mL
PROOVHG-PCRN-500
VHG-PCRN-500
Chromium
Cr
1000 mg/L
5% HNO₃
500 mL
1 * 500 mL
-
+
1 * 500 mL
-
+
PROOVHG-TCRN-100
VHG-TCRN-100
Chromium
Cr
10000 mg/L
5% HNO₃
100 mL
1 * 100 mL
-
+
1 * 100 mL
-
+
PROOVHG-TCRN-500
VHG-TCRN-500
Chromium
Cr
10000 mg/L
5% HNO₃
500 mL
1 * 500 mL
-
+
1 * 500 mL
-
+
Blocked
This product has been blocked by your organization. Please contact your purchasing department for more information