Norway
Order Entry
Cleaning agent, TICKOPUR J 80 U
Cleaning agent, TICKOPUR J 80 U
undefined
Cleaning agent, TICKOPUR J 80 U
  •  
    Capacity
    pH value
    Pk
  • TICKOPUR J 80 U
    TICKOPUR J 80 U
    142-1945
    880
    1 l
    2
    1 L
    1 * 1 L
    1 * 1 L - kr 285,00
    kr 285,00
    1 * 1 L
       kr 285,00
    Specifications
    Environmentally Preferable
  • TICKOPUR J 80 U
    TICKOPUR J 80 U
    BAND881
    881
    5 l
    2
    5 L
    1 * 5 L
    1 * 5 L - kr 815,00
    kr 815,00
    1 * 5 L
       kr 815,00
    Specifications
    Environmentally Preferable