Norway
Order Entry
Single channel pipettes, mechanical, fixed / variable volume, Research® plus (General Lab Product)
Single channel pipettes, mechanical, fixed / variable volume, Research® plus (General Lab Product)
Supplier:  EPPENDORF
   Sustainable Options Available
Certificates
undefined
Single channel pipettes, mechanical, fixed / variable volume, Research® plus (General Lab Product)
Supplier:  EPPENDORF
   Sustainable Options Available
Certificates
Specifications
  • Catalog No:
  • 613-5902
  • 613-5896
  • 613-6973
  • 613-5895
  • 613-5897
  • 613-5898
  • 613-5900
  • EPPE3124000.113
  • EPPE3124000.121
  • 613-5893
  • 613-5894
  • 613-5901
  • EPPE3124000.016
  • EPPE3124000.105
  • EPPE3124000.040
  • EPPE3124000.059
  • EPPE3124000.067
  • EPPE3124000.075
  • 613-5899
  • EPPE3124000.024
  • EPPE3124000.032
  • EPPE3124000.083
  • 613-7085
  • EPPE3124000.091
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Specifications
Related Information
Variable volume
Product Information
  • Catalog #
    Supplier #
    Volume
    Accuracy
    Imprecision
    Colour code
    Recommended tips
    Availability
    Price
  • 613-5893
    3123000.012
    0,1 - 2,5 µl
    0,1 µl: ±48 % / 0,048 µl
    0,25 µl: ±12 % / 0,03 µl
    1,25 µl: ±2,5 % / 0,031 µl
    2,5 µl: ±1,4 % / 0,035 µl
    0,1 µl: ±12 % / 0,012 µl
    0,25 µl: ±6,0 % / 0,015 µl
    1,25 µl: ±1,5 % / 0,019 µl
    2,5 µl: ±0,7 % / 0,018 µl
    Dark grey
    epT.I.P.S.® 10 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 975,00
    kr 3 975,00
    1 * 1 Items
       kr 3 975,00
    Supplier Article Number: 3123000.012
    Local Article Number: EPPE3123000.012
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 613-6973
    3123000.144
    0,25 - 2,5 ml
    250 µl: ±4,8 % / 12 µl
    1250 µl: ±0,8 % / 10 µl
    2500 µl: ±0,6 % / 15 µl
    250 µl: ±1,2 % / 3 µl
    1250 µl: ±0,2 % / 2,5 µl
    2500 µl: ±0,2 % / 5 µl
    Red
    epT.I.P.S.® 2,5 ml µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 975,00
    kr 3 975,00
    1 * 1 Items
       kr 3 975,00
    Supplier Article Number: 3123000.144
    Local Article Number: EPPE3123000.144
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 613-5894
    3123000.020
    0,5 - 10 µl
    0,5 µl: ±8,0 % / 0,04 µl
    1 µl: ±2,5 % / 0,025 µl
    5 µl: ±1,5 % / 0,075 µl
    10 µl: ±1,0 % / 0,1 µl
    0,5 µl: ±5,0 % / 0,025 µl
    1 µl: ±1,8 % / 0,018 µl
    5 µl: ±0,8 % / 0,004 µl
    10 µl: ±0,4 % / 0,04 µl
    Grey
    epT.I.P.S.® 20 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 975,00
    kr 3 975,00
    1 * 1 Items
       kr 3 975,00
    Supplier Article Number: 3123000.020
    Local Article Number: EPPE3123000.020
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 613-5895
    3123000.039
    2 - 20 µl
    2 µl: ±5,0 % / 0,1 µl
    10 µl: ±1,2 % / 0,12 µl
    20 µl: ±1,0 % / 0,2 µl
    2 µl: ±1,5 % / 0,03 µl
    10 µl: ±0,6 % / 0,06 µl
    20 µl: ±0,3 % / 0,06 µl
    Yellow
    epT.I.P.S.® 200 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 975,00
    kr 3 975,00
    1 * 1 Items
       kr 3 975,00
    Supplier Article Number: 3123000.039
    Local Article Number: EPPE3123000.039
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 613-5896
    3123000.098
    2 - 20 µl
    2 µl: ±5,0 % / 0,1 µl
    10 µl: ±1,2 % / 0,12 µl
    20 µl: ±1,0 % / 0,2 µl
    2 µl: ±1,5 % / 0,03 µl
    10 µl: ±0,6 % / 0,06 µl
    20 µl: ±0,3 % / 0,06 µl
    Light grey
    epT.I.P.S.® 20 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 975,00
    kr 3 975,00
    1 * 1 Items
       kr 3 975,00
    Supplier Article Number: 3123000.098
    Local Article Number: EPPE3123000.098
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 613-5897
    3123000.047
    10 - 100 µl
    10 µl: ±3,0 % / 0,3 µl
    50 µl: ±1,0 % / 0,5 µl
    100 µl: ±0,8 % / 0,8 µl
    10 µl: ±1,0 % / 0,1 µl
    50 µl: ±0,3 % / 0,15 µl
    100 µl: ±0,2 % / 0,2 µl
    Yellow
    epT.I.P.S.® 200 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 975,00
    kr 3 975,00
    1 * 1 Items
       kr 3 975,00
    Supplier Article Number: 3123000.047
    Local Article Number: EPPE3123000.047
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 613-5898
    3123000.055
    20 - 200 µl
    20 µl: ±2,5 % / 0,5 µl
    100 µl: ±1,0 % / 1,0 µl
    200 µl: ±0,6 % / 1,2 µl
    20 µl: ±0,7 % / 0,14 µl
    100 µl: ±0,3 % / 0,3 µl
    200 µl: ±0,2 % / 0,4 µl
    Yellow
    epT.I.P.S.® 200 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 975,00
    kr 3 975,00
    1 * 1 Items
       kr 3 975,00
    Supplier Article Number: 3123000.055
    Local Article Number: EPPE3123000.055
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 613-5899
    3123000.101
    30 - 300 µl
    30 µl: ±2,5 % / 0,75 µl
    150 µl: ±1,0 % / 1,5 µl
    300 µl: ±0,6 % / 1,8 µl
    30 µl: ±0,7 % / 0,21 µl
    150 µl: ±0,3 % / 0,45 µl
    300 µl: ±0,2 % / 0,6 µl
    Orange
    epT.I.P.S.® 300 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 975,00
    kr 3 975,00
    1 * 1 Items
       kr 3 975,00
    Supplier Article Number: 3123000.101
    Local Article Number: EPPE3123000.101
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 613-5900
    3123000.063
    100 - 1000 µl
    100 µl: ±3,0 % / 3,0 µl
    500 µl: ±1,0 % / 5,0 µl
    1000 µl: ±0,6 % / 6,0 µl
    100 µl: ±0,6 % / 0,6 µl
    500 µl: ±0,2 % / 1,0 µl
    1000 µl: ±0,2 % / 2,0 µl
    Blue
    epT.I.P.S.® 1000 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 975,00
    kr 3 975,00
    1 * 1 Items
       kr 3 975,00
    Supplier Article Number: 3123000.063
    Local Article Number: EPPE3123000.063
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 613-5901
    3123000.071
    500 - 5000 µl
    500 µl: ±2,4 % / 12 µl
    2500 µl: ±1,2 % / 30 µl
    5000 µl: ±0,6 % / 30 µl
    500 µl: ±0,6 % / 3 µl
    2500 µl: ±0,25 % / 6,25 µl
    5000 µl: ±0,15 % / 7,5 µl
    Purple
    epT.I.P.S.® 5000 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 975,00
    kr 3 975,00
    1 * 1 Items
       kr 3 975,00
    Supplier Article Number: 3123000.071
    Local Article Number: EPPE3123000.071
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 613-5902
    3123000.080
    1000 - 10000 µl
    1000 µl: ±3,0 % / 30 µl
    5000 µl: ±0,8 % / 40 µl
    10000 µl: ±0,6 % / 60 µl
    1000 µl: ±0,6 % / 6 µl
    5000 µl: ±0,2 % / 10 µl
    10000 µl: ±0,15 % / 15 µl
    Turquoise
    epT.I.P.S.® 10000 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 975,00
    kr 3 975,00
    1 * 1 Items
       kr 3 975,00
    Supplier Article Number: 3123000.080
    Local Article Number: EPPE3123000.080
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
Fixed volume
Product Information
  • Catalog #
    Supplier #
    Volume
    Accuracy
    Imprecision
    Colour code
    Recommended tips
    Availability
    Price
  • EPPE3124000.024
    3124000.024
    10 µl
    ±1,2 %
    ±0,6 %
    Yellow
    epT.I.P.S.® 200 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 475,00
    kr 2 475,00
    1 * 1 Items
       kr 2 475,00
    Supplier Article Number: 3124000.024
    Local Article Number: EPPE3124000.024
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • EPPE3124000.032
    3124000.032
    20 µl
    ±0,8 %
    ±0,3 %
    Light grey
    epT.I.P.S.® 20 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 475,00
    kr 2 475,00
    1 * 1 Items
       kr 2 475,00
    Supplier Article Number: 3124000.032
    Local Article Number: EPPE3124000.032
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • EPPE3124000.040
    3124000.040
    20 µl
    ±1,0 %
    ±0,3 %
    Yellow
    epT.I.P.S.® 200 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 475,00
    kr 2 475,00
    1 * 1 Items
       kr 2 475,00
    Supplier Article Number: 3124000.040
    Local Article Number: EPPE3124000.040
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • EPPE3124000.067
    3124000.067
    50 µl
    ±0,7 %
    ±0,3 %
    Yellow
    epT.I.P.S.® 200 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 475,00
    kr 2 475,00
    1 * 1 Items
       kr 2 475,00
    Supplier Article Number: 3124000.067
    Local Article Number: EPPE3124000.067
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • EPPE3124000.091
    3124000.091
    200 µl
    ±0,6 %
    ±0,2 %
    Blue
    epT.I.P.S.® 1000 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 475,00
    kr 2 475,00
    1 * 1 Items
       kr 2 475,00
    Supplier Article Number: 3124000.091
    Local Article Number: EPPE3124000.091
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • EPPE3124000.113
    3124000.113
    500 µl
    ±0,6 %
    ±0,2 %
    Blue
    epT.I.P.S.® 1000 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 475,00
    kr 2 475,00
    1 * 1 Items
       kr 2 475,00
    Supplier Article Number: 3124000.113
    Local Article Number: EPPE3124000.113
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • EPPE3124000.121
    3124000.121
    1000 µl
    ±0,6 %
    ±0,2 %
    Blue
    epT.I.P.S.® 1000 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 475,00
    kr 2 475,00
    1 * 1 Items
       kr 2 475,00
    Supplier Article Number: 3124000.121
    Local Article Number: EPPE3124000.121
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • EPPE3124000.016
    3124000.016
    10 µl
    ±1,2 %
    ±0,6 %
    Grey marl
    epT.I.P.S.® 20 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 475,00
    kr 2 475,00
    1 * 1 Items
       kr 2 475,00
    Supplier Article Number: 3124000.016
    Local Article Number: EPPE3124000.016
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • EPPE3124000.059
    3124000.059
    25 µl
    ±1,0 %
    ±0,3 %
    Yellow
    epT.I.P.S.® 200 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 475,00
    kr 2 475,00
    1 * 1 Items
       kr 2 475,00
    Supplier Article Number: 3124000.059
    Local Article Number: EPPE3124000.059
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • EPPE3124000.075
    3124000.075
    100 µl
    ±0,6 %
    ±0,2 %
    Yellow
    epT.I.P.S.® 200 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 475,00
    kr 2 475,00
    1 * 1 Items
       kr 2 475,00
    Supplier Article Number: 3124000.075
    Local Article Number: EPPE3124000.075
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • EPPE3124000.083
    3124000.083
    200 µl
    ±0,6 %
    ±0,2 %
    Yellow
    epT.I.P.S.® 200 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 475,00
    kr 2 475,00
    1 * 1 Items
       kr 2 475,00
    Supplier Article Number: 3124000.083
    Local Article Number: EPPE3124000.083
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • EPPE3124000.105
    3124000.105
    250
    ±0,6 %
    ±0,2 %
    Blue
    epT.I.P.S.® 1000 µl
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 475,00
    kr 2 475,00
    1 * 1 Items
       kr 2 475,00
    Supplier Article Number: 3124000.105
    Local Article Number: EPPE3124000.105
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
6-Packs
Product Information
  • Catalog #
    Supplier #
    Volume
    Accuracy
    Imprecision
    Colour code
    Recommended tips
    Availability
    Price
  • Eppendorf Research® plus, 6-pack with pipette carousel 2, 1-channel, variable, including 6× epT.I.P.S.® box 2.0 and ballpoint pen, 0,1 - 2,5 µl, 0,5 - 10 µl, 2 - 20 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl
    Eppendorf Research® plus, 6-pack with pipette carousel 2, 1-channel, variable, including 6× epT.I.P.S.® box 2.0 and ballpoint pen, 0,1 - 2,5 µl, 0,5 - 10 µl, 2 - 20 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl
    613-7085
    3123000.942
    Yellow
    -
    Log in for availability
    1 * 1 Each
    1 * 1 Each - kr 20 400,00
    kr 20 400,00
    1 * 1 Each
       kr 20 400,00
    Supplier Article Number: 3123000.942
    Local Article Number: EPPE3123000.942
    Specifications:
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
3-Packs
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 01
    Option 1, incl. 3 Eppendorf Research® plus pipettes (0,5 - 10 µl, 10 - 100 µl, 100 - 1000 µl) and epT.I.P.S.® boxes
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - kr 9 725,00
    kr 9 725,00
    Supplier Article Number: 3123000.900
    Local Article Number: EPPE3123000.900
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 02
    Option 2, incl. 3 Eppendorf Research® plus pipettes (2 - 20 µl, 20 - 200 µl, 100 - 1000 µl) and epT.I.P.S.® boxes
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - kr 9 725,00
    kr 9 725,00
    Supplier Article Number: 3123000.918
    Local Article Number: EPPE3123000.918
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 03
    Option 3, incl. 3 Eppendorf Research® plus pipettes (100 - 1000 µl, 0,5 - 5 ml, 1 - 10 ml) and epT.I.P.S.® samples
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - kr 9 725,00
    kr 9 725,00
    Supplier Article Number: 3123000.926
    Local Article Number: EPPE3123000.926