Vydac 238MS C18, HPLC columns
Supplier: Avantor
Certificates
About this item
These Vydac MS columns are a later development from Grace for the reversed phase HPLC separation of biomolecules. A proprietary surface treatment and bonding process gives Vydac MS columns a unique selectivity. A variety of reversed phases makes this product line suitable for the analysis of small peptides to large intact, undenatured proteins.
- Five reversed-phase chemistries
- Excellent peak shape with little or no TFA
- High protein recoveries make scale-up easy
Monomeric bonding offers increased peptide interaction and generally yields higher peak counts. Use for same applications as 218MS, but offers different C18 selectivity.
Vydac is a registered trademark of W.R.Grace and Co.
Specifications
- Catalog No:
- 238GK54MS/N
- 238MS5.310
- Environmentally Preferable:
- Low Manufacturing Impact
- Carbon load:
- 4%
- Endcapped:
- Yes
- USP code:
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
Specifications
Product Information
- Catalog #Column FormatPhaseColumn hardwarePore sizeParticle SizeØ int.AvailabilityPrice
- 238MS5.310CapillaryC18Stainless steel300 Å5 µm0,30 mmLog in for availability1 * 1 Itemskr 13 000,00Local Article Number: HICH238MS5.310Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYesUSP codeL1Environmental certification
Guard cartridges
Product Information
- Catalog #Column FormatPhaseColumn hardwarePore sizeParticle SizeØ int.AvailabilityPrice
- Guard kit for 3,0 – 4,6 mm ID columns, includes 1× guard cartridge, holder and couplerGuard kit for 3,0 – 4,6 mm ID columns, includes 1× guard cartridge, holder and coupler238GK54MS/NGuard cartridgeC18Stainless steel5 µmLog in for availability1 * 1 Itemskr 5 550,00Local Article Number: HICH238GK54MS/NSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact