Spigot, stainless steel
Leverandør: Bürkle
BURK0535-0020EA 5300 NOK
BURK0535-0020
Spigot, stainless steel
Haner
It is Very sturdy and robust spigot made of stainless steel. The outlet of the spigot turns 360° so it can be adjusted to any required position.
  • Seals PTFE, FKM
  • Self-closing
  • Lockable
  • NW 10 mm