ESD safety shoes, sandals, Velcro®, Basic Comfort
Leverandør: Honeywell Safety Products
Basic Comfort
DALL98445-625/42EA 1550 NOK
DALL98445-625/42 DALL98445-625/36 DALL98445-625/44 DALL98445-625/35 DALL98445-625/47 DALL98445-625/43 DALL98445-625/41 DALL98445-625/38 DALL98445-625/37 DALL98445-625/40 DALL98445-625/45 DALL98445-625/46 DALL98445-625/39 DALL98445-625/48
ESD safety shoes, sandals, Velcro®, Basic Comfort
Sko og støvler
ESD safety shoes made from water repellent nubuck and suede leather upper plus a durable, anti-slip, dual-density TPU/PU sole, resistant to oil and fuel.
  • Premium textile lining
  • Orthopaedic insole possible
  • Light density, shock-absorbent PU midsole
  • Protective steel toecap
Width: 9 (35-37); width 11 (38-48)
Colour: Black/grey
Sertifiseringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1, SRC