Z-Leu-Leu-Glu-Amc
Catalog # ENZOBMLZW93450005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:02
- Protein/peptide name:Z-Leu-Leu-Glu-AMC
Specifications
About this item
Fluorogenic substrate for the peptidylglutamyl-peptide hydrolysing (caspase-like) activity of the proteasome which may be stimulated in the presence of Mg2+ ions and rapidly inactivated by N-acetylimidazole.