Norway
Order Entry
Z-Leu-Leu-Glu-Amc
Z-Leu-Leu-Glu-Amc
Catalog # ENZOBMLZW93450005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Z-Leu-Leu-Glu-Amc
Catalog # ENZOBMLZW93450005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Price
kr 2 025,00
Log in for availability
Unit of Measure 1 * 5 mg
Specifications
  • Pk:
    5 mg
  • Seq:
    02
  • Protein/peptide name:
    Z-Leu-Leu-Glu-AMC
Specifications
About this item
Fluorogenic substrate for the peptidylglutamyl-peptide hydrolysing (caspase-like) activity of the proteasome which may be stimulated in the presence of Mg2+ ions and rapidly inactivated by N-acetylimidazole.