High visibility waistcoat, CL 2 501 H
Leverandør: Fristads Kansas Group
KWIN100382130M/LEA 200 NOK
KWIN100382130M/L KWIN100382130XL2XL KWIN100382130XS/S KWIN1003821303XL
High visibility waistcoat, CL 2 501 H
Arbeidstøy
Waistcoat with two reflective strips, made from 100% polyester.
  • Velcro fastening
  • Washable at up to 40 °C
Weight: 130 g/m²
Sertifiseringer: EN ISO 20471 (class 1 - size XS/S and class 2 - size M/L-3XL/4XL)