Pyruvate oxidase
550701-100
:
- Pk:100 EU
- CAS No.:9001-96-1
- Enzyme name:Pyruvate oxidase
- Enzyme activity:≥25 U/mg
- pH:6,5
Pyruvate oxidase from Aerococcus viridans catalyses the oxidation of pyruvate to acetyl phosphate, carbon dioxide, and hydrogen peroxide in the presence of phosphate.
- Lyophilized yellow solid