Caps for test/culture tubes without rim
: LUDI
Made of anodised aluminium.
- Springs clamp against the outer wall of the vessel
- Anodised and coloured
- Autoclavable
Without grip. Cap for sterile but not hermetic closing of test tubes and other vessels.
- For Ø ext.ColourPk
- LUDI18401063118401063115 - 16 mmSilver1001 * 100 Itemskr 4 100,00
Specifications
Environmentally Preferable - LUDI18401074118401074117 - 18 mmSilver1001 * 100 Itemskr 4 350,00
Specifications
Environmentally Preferable - LUDI18401074218401074217 - 18 mmRed1001 * 100 Itemskr 4 350,00
Specifications
Environmentally Preferable - LUDI18401074318401074317 - 18 mmBlue1001 * 100 Itemskr 4 350,00
Specifications
Environmentally Preferable - LUDI18401074418401074417 - 18 mmYellow1001 * 100 Itemskr 4 350,00
Specifications
Environmentally Preferable - LUDI18401074518401074517 - 18 mmGreen1001 * 100 Itemskr 4 375,00
Specifications
Environmentally Preferable