ESD occupational shoes, clogs, Easy, 37620
Leverandør: Abeba
Easy
ABEB37620/36EA 1100 NOK
ABEB37620/36 ABEB37620/37 ABEB37620/38 ABEB37620/39 ABEB37620/40 ABEB37620/41 ABEB37620/42 ABEB37620/43 ABEB37620/35
ESD occupational shoes, clogs, Easy, 37620
Sko og støvler
ESD clog made from microfibre with a non foldable, adjustable heel strap. The TPU sole is resistant to fuel, oil and animal fats.
  • Replaceable acc Wave insoles
  • Sanitized® treatment reduces odour and bacterial growth
  • TPU sole with high slip resistance
Suitable for use in kitchens and in food processing industries.
Colour: White
Sertifiseringer: EN 61340 (ESD); EN ISO 20347:2012; OB, FO, E, A, SRC