Robotic tips for Beckman Multimek Type II, Axygen®
Leverandør: Corning
Axygen®
AXYGRMT-200-BK-REA 10000 NOK
AXYGRMT-200-BK-R 736-0470
Robotic tips for Beckman Multimek Type II, Axygen®
Pipettespisser
Robbins-style tips for Beckman Multimek 'Type II' systems.
  • Accuracy and precision testing is performed for every manufacturing lot using the finest automated systems in the industry
  • Certified RNase-, DNase-, and endotoxin-free
  • Available in black or transparent
  • Non filtered, non sterile
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Available in nonsterile, sterile, non-filtered, and filtered formats. Black and clear polypropylene tips are available in 384-well format designed for Molecular Devices Flexstation systems.