Hank's balanced salt solutions (HBSS), Corning®
: Corning
Hanks' Balanced Salt Solution (HBSS) is a buffer used to maintain a physiological pH for cells maintained in non-CO₂ atmospheric conditions.
- Low endotoxin - < 0.25 EU/ml
- Mycoplasma tested
- Liquid media products are formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
- Mycoplasma:Tested Negative
- Endotoxin content:< 0.25 EU/mL
- Shelf life:36 months
- Storage temperature:2 °C to 8 °C
- Pk InfoPk
- HBSS (Hank’s Balanced Salt Solution) 10X without Sodium bicarbonate, Calcium and MagnesiumHBSS (Hank’s Balanced Salt Solution) 10X without Sodium bicarbonate, Calcium and MagnesiumMDTC20-021-CVR20-021-CVR6×500 ml61 * 6 Itemskr 2 800,00
- HBSS (Hank’s Balanced Salt Solution) 10X with Calcium and Magnesium, without Phenol red and Sodium bicarbonateHBSS (Hank’s Balanced Salt Solution) 10X with Calcium and Magnesium, without Phenol red and Sodium bicarbonate20-023-CV6×500 ml61 * 6 Itemskr 2 825,00
- HBSS (Hank’s Balanced Salt Solution) 1X with Calcium and MagnesiumHBSS (Hank’s Balanced Salt Solution) 1X with Calcium and Magnesium21-020-CV6×500 ml61 * 6 Itemskr 1 350,00
- HBSS (Hank’s Balanced Salt Solution) 1X with Calcium and MagnesiumHBSS (Hank’s Balanced Salt Solution) 1X with Calcium and Magnesium21-020-CM6×1 litre61 * 6 Itemskr 2 250,00
- HBSS (Hank’s Balanced Salt Solution) 1X with Calcium and Magnesium,
without Phenol redHBSS (Hank’s Balanced Salt Solution) 1X with Calcium and Magnesium,
without Phenol red21-023-CV6×500 ml61 * 6 Itemskr 1 575,00 - HBSS (Hank’s Balanced Salt Solution) 1X with Calcium and Magnesium,
without Phenol redHBSS (Hank’s Balanced Salt Solution) 1X with Calcium and Magnesium,
without Phenol red21-023-CM6×1 litre61 * 6 Itemskr 1 375,00 - HBSS (Hank’s Balanced Salt Solution) 1X without Calcium and MagnesiumHBSS (Hank’s Balanced Salt Solution) 1X without Calcium and Magnesium21-021-CV6×500 ml61 * 6 Itemskr 1 550,00
- HBSS (Hank’s Balanced Salt Solution) 1X without Calcium and MagnesiumHBSS (Hank’s Balanced Salt Solution) 1X without Calcium and Magnesium21-021-CM6×1 litre61 * 6 Itemskr 2 250,00
- HBSS (Hank’s Balanced Salt Solution) 1X without Calcium, Magnesium and Phenol redHBSS (Hank’s Balanced Salt Solution) 1X without Calcium, Magnesium and Phenol red21-022-CV6×500 ml61 * 6 Itemskr 1 050,00
- HBSS (Hank’s Balanced Salt Solution) 1X without Calcium, Magnesium and Phenol redHBSS (Hank’s Balanced Salt Solution) 1X without Calcium, Magnesium and Phenol red21-022-CM6×1 litre61 * 6 Itemskr 2 375,00