Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
kr 0,00
Order Entry
Home
DL-Malic acid
DL-Malic acid
DL-Malic acid
DL-Malic acid
EDQMY0000143
:
EDQM
:
DL-Malic acid
EDQMY0000143
:
EDQM
:
kr 3 225,00
1 * 1 Items
This product has been blocked by your organization. Please contact your purchasing department for more information
Pk:
1
Seq:
0001
:
:
134.09 g/mol
:
13,05730
:
306 °C (1013 hPa)
:
131…132 °C
:
1.601 g/cm³ (20 °C)
:
210-514-9
:
GHS05
:
GHS07
:
Ambient
:
617-48-1
:
HOOCCH(OH)CH₂COOH
:
MFCD00064212