Fluorsyre 50%, VLSI Selectipur® for elektronisk industri
|
Danger
|
50488860.EA
1300
NOK
50488860.
Fluorsyre 50%, VLSI Selectipur® for elektronisk industri
Fluorsyre
Product from BASF
NB: This product is not available in all countries. Please check with your local VWR office or supplier.
Formel:
HF MW: 20,01 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00011346 CAS-nummer: 7664-39-3 EINECS: 231-634-8 FN: 1790 ADR: 8 (6.1),II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...