Tiourea

Leverandør: Cytiva
Warning

RPN6301EA 1050 NOK
RPN6301
Tiourea
Tiourea

Thiourea is a chaotrope used together with urea in the IPG strip rehydration to solubilise and denature proteins, and improve the solubility of membrane proteins.

Formel: NH₂CSNH₂
MW: 76,12 g/mol
Kokepunkt: (dec.)
Smeltepunkt: 176…178 °C
Tetthet: 1,435 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008067
CAS-nummer: 62-56-6
EINECS: 200-543-5
FN: 3077
ADR: 9,III
Merck Indeks: 12,09505
https://no.vwr.com/stibo/web/std.lang.all/72/67/2297267.jpg

Bestill nå

Spesifikasjon Testresultater

Application Used together with urea in the IPG strip rehydration solution to solubilize and denature proteins
Color White
Melting/Freezing Point 176-178°C
Molecular Weight (Mr) 76.13
pH Range 5-7
Physical State Solid
Solubility Easily soluble in methanol, soluble in cold and hot water, partially soluble in diethyl ether
Synonym Sulfourea, Thiocarbamide
Wetted Part No

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR