Iodoacetamide, proteomics kvalitet
Leverandør: Cytiva
Warning
RPN6302EA 1250 NOK
RPN6302
Iodoacetamide, proteomics kvalitet
Iodoacetamide
Used for the efficient alkylation of thiols while minimizing oxidation of competing thiol pairs in protein samples.
  • Proteomic grade
  • Easily soluble in cold and hot water
Ultrapure, proteomic-grade iodoacetamide is used for efficient alkylation of thiols while minimizing oxidation of the competing thiol pairs in protein samples.
Formel: ICH₂CONH₂
MW: 184,96 g/mol
Kokepunkt: 266 °C (1013 hPa)
Smeltepunkt: 93…96 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00008028
CAS-nummer: 144-48-9
EINECS: 205-630-1
FN: 2811
ADR: 6.1,III