Du søkte etter: Nucleic Acid Assays
25  treff i ditt søk
SearchResultCount:"25"
Sort Results
Listevisning Easy View (ny)
Vurder dette søkeresultatet
Artikkenummer: (1B1558-KIT)
Produsent: VWR Chemicals
Beskrivelse: Fluorescent DNA Quantitation Kit uses fluorescent bisBenzimide H33258 dye to bind to AT sequences of the minor groove of double-stranded DNA. In the a...
UOM: 1 * 1 Each
Produsent: Biotium
Beskrivelse: AccuBlue® and AccuClear® dsDNA Quantitation Kits offer sensitive and selective detection of double-stranded DNA in a 96-well microplate format.
HMS
Produsent: Oxford Nanopore Technologies
Beskrivelse: MinION™ ― your personal, portable DNA and RNA sequencer.
Produsent: Quantabio
Beskrivelse: Ultra FAST RNA library prep with integrated rRNA and globin depletion.
Produsent: Oxford Nanopore Technologies
Beskrivelse: MinION™ Mk1C ― your 'all in one', portable DNA and RNA sequencer.
Artikkenummer: (AATB17589)
Produsent: AAT BIOQUEST INC
Beskrivelse: Gelite™ Green is a sensitive fluorescent nucleic acid gel stain for detecting nucleic acids in agarose and polyacrylamide gels.
UOM: 1 * 1 Each
New Product
Produsent: AAT BIOQUEST INC
Beskrivelse: DNA Quantitation is a very important task in DNA sample preparations for various genomic analyses.
New Product
Produsent: Biotium
Beskrivelse: The AccuBlue® broad range RNA quantitation kit boasts the widest linear range (5 to 1000 ng RNA) of available RNA quantitation kits, while providing e...
HMS
Produsent: AAT BIOQUEST INC
Beskrivelse: DNA Quantitation is a very important task in DNA sample preparations for various analyses.
New Product
Artikkenummer: (AATB17594)
Produsent: AAT BIOQUEST INC
Beskrivelse: Gelite™ Orange is an extremely sensitive nucleic acid gel stain for detecting DNA or RNA in gels using a standard 300 nm UV transilluminator and Polar...
UOM: 1 * 1 Each
New Product
Artikkenummer: (AATB17651)
Produsent: AAT BIOQUEST INC
Beskrivelse: Helixyte™ Green dsDNA Quantitation Assay Kit can be used for selectively detecting as little as 25 pg/ml of dsDNA in the presence of ssDNA, RNA, and f...
UOM: 1 * 1 Each
New Product
Artikkenummer: (AATB17656)
Produsent: AAT BIOQUEST INC
Beskrivelse: Detecting and quantitating small amounts of RNA is extremely important for a wide variety of molecular biology procedures such as measuring yields of ...
UOM: 1 * 1 Each
New Product
Artikkenummer: (AATB17631)
Produsent: AAT BIOQUEST INC
Beskrivelse: Portelite™ fluorimetric total nucleic acid quantitation kit is optimised for measuring the total amounts of nucleic acids with CytoCite™ or Qubit® flu...
UOM: 1 * 1 Each
New Product
Artikkenummer: (AATB17623)
Produsent: AAT BIOQUEST INC
Beskrivelse: Helixyte™ Green fluorimetric ssDNA quantitation kit is optimised for quantifying dsDNA with a fluorescence microplate reader.
UOM: 1 * 1 Each
New Product
Produsent: AAT BIOQUEST INC
Beskrivelse: Detecting and quantitating small amounts of RNA is extremely important for a wide variety of biological applications such as measuring yields of in vi...
New Product
Artikkenummer: (AATB17630)
Produsent: AAT BIOQUEST INC
Beskrivelse: Helixyte™ Green fluorimetric total nucleic acid quantitation kit is optimised for measuring the total amounts of nucleic acids with a fluorescence mic...
UOM: 1 * 1 Each
New Product
Pris på forespørsel
Lager for dette produktet er begrenset, men kan være tilgjengelig i en lagerbygning i nærheten av deg. Vennligst sørg for at du er logget inn på nettstedet, slik at tilgjengelige lager kan vises. Hvis call er fortsatt vises og du trenger hjelp, kan du ringe oss på 1-800-932 - 5000.
Lager for dette punktet er begrenset, men kan være tilgjengelig i en lagerbygning i nærheten av deg. Vennligst sørg for at du er logget inn på nettstedet, slik at tilgjengelige lager kan vises. Hvis call er fortsatt vises og du trenger hjelp, kan du ringe oss på 1-800-932 - 5000.
NB Dette produkt er regulert i henhold til norsk lov.
Dersom ytterligere informasjon er nødvendig fra deg vil du bli kontaktet av VWR via e-post.
OBS! Etanol er underlagt bestemmelse om særavgift, jfr LOV-1993-05-19-11-§1. VWR selger kun etanol til kunder som kan fremlegge avgiftsfritak for udenaturert etanol, jfr. § 3-3-7 ( forskrifter om særavgifter).
NB Dette produkt er regulert i henhold til norsk lov.
Dersom ytterligere informasjon er nødvendig fra deg vil du bli kontaktet av VWR via e-post.
OBS! Etanol er underlagt bestemmelse om særavgift, jfr LOV-1993-05-19-11-§1. VWR selger kun etanol til kunder som kan fremlegge avgiftsfritak for udenaturert etanol, jfr. § 3-3-7 ( forskrifter om særavgifter).
Dette produktet har blitt blokkert for din organisasjon. Ta kontakt med din innkjøpsavdeling for mer informasjon.
Det opprinnelige produktet er ikke lenger tilgjengelig. Varen som vises er tilgjengelig.
Produkt (er) merket med dette symbolet er ikke lenger tilgjengelig og selges til lageret er tomt. Alternativer kan være tilgjengelige ved å søke med VWR Katalognummer ovenfor. Hvis du trenger ytterligere hjelp, ring VWR kundeservice 22900000
1 - 16 of 25
no targeter for Bottom