
Barium diphenylamine-4-sulphonate, Millipore®
1.00255.0005
: Merck
:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:300 °C
- Molar Mass:633.88 g/mol
- Bulk Density:220 kg/m3
- pH:5.9 (10 g/l, H2O, 20 °C) (slurry)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
:
:633.89 g/mol
:300 °C
:Ambient
:1564
:6.1,III
:M-Clarity™ quality level = MQ200
:228-278-0
:GHS07
:6211-24-1
:1300-92-1
:C₂₄H₂₀BaN₂O₆S₂
:MFCD00007497