Decafluorobensofenon ≥98%
Leverandør: Thermo Scientific
A19262.06EA
2250
NOK
A19262.06
A19262.03
Decafluorobensofenon ≥98%
Decafluorobensofenon
Formel:
C₁₃F₁₀O MW: 362,13 g/mol Kokepunkt: 263 °C (1013 hPa) Smeltepunkt: 92…94 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00000295 CAS-nummer: 853-39-4 EINECS: 212-717-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...