Norway
Order Entry
Isoniazid
  PROOICRS0331
 :  
Isoniazid
  PROOICRS0331
 :  
  1 * 100 mg
  • Pakketype:
    Glass bottle for solids
  • Fp:
    100 mg
  • Seq:
    2440
 :
 :137.14 g/mol
 :13,05205
 :169…172 °C
 :1,42 g/cm³ (20 °C)
 :>250 °C
 :200-214-6
 :GHS07
 :Romtemperatur
 :54-85-3
 :C₆H₇N₃O
 :MFCD00006426