Buckets, UN approved, tinplate
Leverandør: Rixius
RIXI17-1300-100230EA 3525 NOK
RIXI17-1300-100230
Buckets, UN approved, tinplate
Made of tinplate, with metal lid.
  • Good temperature resistance
  • With clamping lock and lid
  • UN approved
For use in laboratories and industry.