Tubing, silicone, TuFlux® SIL
Leverandør: Sartorius
TuFlux® SIL
SARPFSA117429
Tubing, silicone, TuFlux® SIL
Slanger Overføringsslange
Designed to facilitate fluid transfer in the biopharmaceutical manufacturing processes.
  • Highly resistant platinum cured silicone tubing
  • Media and buffer processing
  • Filtration and fermentation
  • Cell harvest
Gamma irradiated and autoclavable.
Temperature resistance: −60 to +200 °C
Hardness (Shore durometer A): 60 ±5