Natriumtiosulfat løsning 0.05 mol/l (0.05 N), Titripur®, Supelco®
M-Clarity™ quality level = MQ100
1.60311.1000EA
331
NOK
1.60311.1000
Natriumtiosulfat løsning 0.05 mol/l (0.05 N), Titripur®, Supelco®
Natriumtiosulfat
Formel:
Na₂S₂O₃ MW: 158,11 g/mol |
MDL nummer:
MFCD00003499 CAS-nummer: 7772-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...