Norway
Order Entry
Hand washing foam, Phago´derm Sensitive
Hand washing foam, Phago´derm Sensitive
Supplier:  PHAGOGENE
Certificates
Hand washing foam, Phago´derm Sensitive
Supplier:  PHAGOGENE
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Hand washing foam, Phago´derm sensitive, 1000 ml pump bottle
    Hand washing foam, Phago´derm sensitive, 1000 ml pump bottle
    PHAG65085
    65085
    12
    Log in for availability
    Log in for availability
    1 * 12 Items
    1 * 12 Items - kr 2 650,00
    kr 2 650,00
    1 * 12 Items
       kr 2 650,00
    Supplier Article Number: 65085
    Local Article Number: PHAG65085
    Specifications:
    Specifications
    Environmentally Preferable
  • Hand washing foam, Phago´derm sensitive, 5 l bottle
    Hand washing foam, Phago´derm sensitive, 5 l bottle
    PHAG65083
    65083
    2
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 720,00
    kr 720,00
    1 * 2 Items
       kr 720,00
    Supplier Article Number: 65083
    Local Article Number: PHAG65083
    Specifications:
    Specifications
    Environmentally Preferable