Gas syringes, series A-2, Pressure-Lok®
: VICI
The syringes feature a PTFE plunger tip, stress-formed by a special process to ensure a leak-tight seal. The self-lubricating plunger tip stays smooth for the life of the syringe, with none of the seizing or residue buildup associated with conventional all-metal plungers.
- Removable needles
- PTFE plunger tip
- PTFE-sealed needle
- Low dead volume
- Crisp clean graduations
Small liquid samples with low-boiling components are not lost through evaporation, as often occurs with ordinary syringes. The positive rear stop prevents plunger blowout at elevated pressures.
- Catalog No:
- VICIPS-050025
- VICIPS-050034
- VICIPS-050035
- VICIPS-050032
- VICIPS-050031
- Series:
- A-2
- A-2
- A-2
- A-2
- A-2
- Max. pressure:
- 250 psi
- 250 psi
- 250 psi
- 250 psi
- 250 psi
- Needle length:
- 0,028×0,005×2"
- 0,029×0,012×2"
- 0,029×0,012×2"
- 0,029×0,012×2"
- 0,029×0,012×2"
Product Information
- Catalog #Supplier #VolumeTypePkAvailabilityPrice
- Gas syringes series A-2Gas syringes series A-2VICIPS-050025PS-050025100 µlStandard11 * 1 Itemskr 2 275,00: PS-050025: VICIPS-050025
Specifications
Environmentally PreferableSeriesA-2Max. pressure250 psiNeedle length0,028×0,005×2" - Gas syringes series A-2: PS-050031: VICIPS-050031
Specifications
Environmentally PreferableSeriesA-2Max. pressure250 psiNeedle length0,029×0,012×2" - Gas syringes series A-2Gas syringes series A-2VICIPS-050032PS-050032500 µlStandard11 * 1 Itemskr 2 025,00: PS-050032: VICIPS-050032
Specifications
Environmentally PreferableSeriesA-2Max. pressure250 psiNeedle length0,029×0,012×2" - Gas syringes series A-2Gas syringes series A-2VICIPS-050034PS-0500342 mlStandard11 * 1 Itemskr 1 650,00: PS-050034: VICIPS-050034
Specifications
Environmentally PreferableSeriesA-2Max. pressure250 psiNeedle length0,029×0,012×2" - Gas syringes series A-2: PS-050035: VICIPS-050035
Specifications
Environmentally PreferableSeriesA-2Max. pressure250 psiNeedle length0,029×0,012×2"