Derivatisation reagents for GC
Leverandør: Macherey-Nagel
MANA701953EA 2575 NOK
MANA701953 MANA701951 MANA701952 MANA701950
Derivatisation reagents for GC
Kromatografiabsorbenter GC sorbenter
The purpose of derivatisation in GC includes:
  • Improvement of volatility, higher thermal stability or a lower detection limit
  • Quantitative, fast and reproducible formation of only one derivative
  • Introduction of halogen atoms for high sensitive ECD detection
  • Influence on elution orders and fragmentation patterns in MS
Reagents for silylation, alkylation (methylation) and acylation are available.
With the following kits a selection of the best suitable derivatisation method and reagent is easy.