Norway
Order Entry
Nitric acid 64 - 66%, Puriss., Fluka™
Nitric acid 64 - 66%, Puriss., Fluka™
  • Assay:
    64 - 66 %
  • Residue on ignition (SO₄):
    Max. 0.0005 %
  • Arsenic (As):
    Max. 0.0001 %
  • Calcium (Ca):
    Max. 0.0005 %
  • Iron (Fe):
    Max. 0.0001 %
  • Vapor Pressure:
    8 mmHg ( 20 °C)
  • Heavy metals (as Pb):
    Max. 0.0005 %
  • Chloride (Cl):
    Max. 0.0001 %
  • Sulfate (SO₄):
    Max. 0.0002 %
  •  
    Pack type
    Pk
  • 07006-1L
    07006-1L
    Plastic bottle
    1 L
    1 * 1 L
    1 * 1 L - kr 640,00
    kr 640,00
    1 * 1 L
       kr 640,00
     : 07006-1L
     : HONC07006-1L
  • 07006-2.5L
    07006-2.5L
    Plastic bottle
    2,5 L
    1 * 2,5 L
    1 * 2,5 L - kr 1 425,00
    kr 1 425,00
    1 * 2,5 L
       kr 1 425,00
     : 07006-2.5L
     : HONC07006-2.5L