Organic reference standard, neat, Fluorene, (CRM), TraceCERT®, Supelco®
Leverandør: Merck
TraceCERT®
56849-100MGEA 700 NOK
56849-100MG
Organic reference standard, neat, Fluorene, (CRM), TraceCERT®, Supelco®
Standarder Organiske standarder
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34).
Forpakning: Bottomless glass bottle. Contents are inside inserted fused cone.