Norway
Order Entry
Ammonia solution 25%, LiChropur™ for HPLC, Supelco®
Corrosive
Irritant
Environment
Ammonia solution 25%, LiChropur™ for HPLC, Supelco®
Supplier:  Merck
Certificates
Corrosive
Irritant
Environment
Ammonia solution 25%, LiChropur™ for HPLC, Supelco®
Supplier:  Merck
Certificates
Related Information
Certificates
Search by Lot Number
Product Information
  • Catalog #
    Supplier #
    Pack type
    Pk
    Availability
    Price
  • 5.43830.0100
    5.43830.0100
    Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - kr 410,00
    kr 410,00
    1 * 100 mL
       kr 410,00
  • 5.43830.0250
    5.43830.0250
    Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - kr 965,00
    kr 965,00
    1 * 250 mL
       kr 965,00