Muffle furnaces with fold down door, Tmax 1300 °C and controller C550, L 5/13 - L 15/13
Leverandør: Nabertherm
Nabertherm
455-1573EA 0 NOK
455-1573 455-1583 455-1363 455-1575 455-1157 455-1574 455-1368 455-1158 455-1369 455-1160 455-1580 455-1151 455-1371 455-1150 455-1362 455-1152 455-1361 455-1581
Muffle furnaces with fold down door, Tmax 1300 °C and controller C550, L 5/13 - L 15/13
Ovner Muffelovner
These furnaces feature heating elements on supporting tubes that radiate freely into the chamber, ensuring particularly short heating times. With the robust lightweight refractory brick insulation, they can reach a max. working temperature of 1300 °C.
  • Heating from two sides
  • Multi-layer insulation with robust lightweight refractory bricks in the furnace chamber
  • Textured stainless steel housing
  • Dual shell housing for low external temperatures and high stability
  • Fold down door (L models) which can be used as a work platform
  • Exhaust air outlet on rear wall of furnace
The furnace is equipped with the C550 standard controller with touch screen display, featuring 10 freely adjustable programs and 20 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. The C550 controllers can automatically activate the vent with fan or catalyst (not model L 15).
Bestillingsinformasjon: All units require a 230 V single-phase power supply. Optional accessories e.g. a vent, vent with fan or catalytic converter, over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, protective gas connection on the rear wall of the furnace. Manual or automatic gas supply systems are also available. Corresponding models with lift up doors, different Tmax and other controllers are also available. Please search the website for more details.
NB: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model.
Note heat up times: If connected at 230 V 1/N/PE rsp. 400 V 3/N/PE.
*Temperature uniformity: ± 5 K with closed air inlet in the empty workspace according to DIN 17052-1 at working temperatures > 800 °C