Chemical resistant sleeve protectors, AlphaTec® 4000, Model 600
Leverandør: Ansell
AlphaTec®
ANSEGR40W-00600-00EA 16300 NOK
ANSEGR40W-00600-00
Chemical resistant sleeve protectors, AlphaTec® 4000, Model 600
Engangsplagg
These sleeve protectors are made from a multi-layer chemical barrier technology that provides an exceptional barrier against a wide range of organic and inorganic chemicals and biological agents.
  • Elasticated at both ends and welded seams
  • Designed to protect
  • Silicone free
Typical applications include: Viral contaminated areas (including Avian Influenza), biological protection, mining, fire and rescue, sewage purification installations.