Muffle furnaces with lift up door, Tmax 1400 °C and controller C550, LT 5/14 - LT 15/14
Leverandør: Nabertherm
Nabertherm
455-1647EA 0 NOK
455-1647 455-1658 455-1449 455-1448 455-1239 455-1649 455-1648 455-1232 455-1242 455-1451 455-1234 455-1454 455-1233 455-1651 455-1453 455-1456 455-1235 455-1653 455-1656 455-1447 455-1237 455-1655 455-1446 455-1240
Muffle furnaces with lift up door, Tmax 1400 °C and controller C550, LT 5/14 - LT 15/14
Ovner Muffelovner
These models stand out for their excellent workmanship, advanced and attractive design, and high level of reliability. Heating elements on support tubes radiating freely into the furnace chamber provide for particularly short heating times and a maximum temperature of 1400 °C. These muffle furnaces are a good alternative to the familiar series when higher application temperatures are needed.
  • Heating from two sides
  • Heating elements on support tubes ensure free heat radiation and a long service life
  • Only fibre materials which are not classified as carcinogenic according to TRGS 905, Class 1 or 2 are used
  • Dual shell housing for low external temperatures and high stability
  • Lift up door (LT) with hot surface facing away from the operator
  • Exhaust air outlet on rear wall of furnace
  • Solid state relays provide low-noise operation
The furnace is equipped with C550 professional controller with touch screen display, featuring 10 freely adjustable programs and 20 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. The C550 controllers can automatically activate the vent with fan or catalytic converter (not model LT 15).
Bestillingsinformasjon: All units require a 230 V single-phase power supply. Optional accessories for example, a vent, vent with fan or catalytic converter, over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, protective gas connection on the rear wall of furnace. Manual or automatic gas supply systems are also available. Corresponding models with fold down doors, different Tmax and other controllers are also available. Please search the website for more details.
NB: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model shown in the table above.
Refence note heat up times: If connected at 230 V 1/N/PE.
* Temperature uniformity of ± 5K in the empty work space for temperatures >800 °C.