UHPLC and HPLC columns, BIOshell™, Supelco®
Leverandør: Merck
BIOshell™
63283-UEA 7375 NOK
63283-U 66915-U 66913-U 66904-U 63288-U 63328-U 66902-U 63325-U
UHPLC and HPLC columns, BIOshell™, Supelco®
Kromatografikolonner
BIOshell™ UHPLC and HPLC columns deliver maximum speed and effciency for the separation of biomolecules on both UHPLC and HPLC systems.
  • Fast and effcient separation of biomolecules
  • Advanced Fused-Core® Technology
  • Pore sizes from 90 up to 1000 Å allows superior separation of glycans as well as very large proteins
The Fused-Core® superficially porous silica particles (SPP) with pore sizes from 90 up to 1000 Å allows superior separation of glycans as well as very large proteins. In particular, a pore size of 1000 Å shows very clear advantages over common 300 Å pores for the separation of very large proteins in biotherapeutic drug development such as monoclonal antibodies (mAbs) or proteins with molecular weights greater than 100 kDa.