ESD safety shoes, lace-up, uvex 2 xenova®, 9568
Leverandør: Uvex
uvex 2 xenova®
UVEX9568.9/52EA 1300 NOK
UVEX9568.9/52 UVEX9568.8/40 UVEX9568.9/51 UVEX9568.7/50 UVEX9568.9/50 UVEX9568.7/39 UVEX9568.7/38 UVEX9568.9/39 UVEX9568.8/49 UVEX9568.8/48 UVEX9568.9/38 UVEX9568.8/47 UVEX9568.8/46 UVEX9568.8/45 UVEX9568.8/44 UVEX9568.8/43 UVEX9568.7/51 UVEX9568.8/42 UVEX9568.7/52 UVEX9568.8/41 UVEX9568.9/41 UVEX9568.8/51 UVEX9568.9/40 UVEX9568.8/50 UVEX9568.7/48 UVEX9568.8/39 UVEX9568.8/38 UVEX9568.7/49 UVEX9568.9/49 UVEX9568.7/46 UVEX9568.9/48 UVEX9568.7/47 UVEX9568.7/44 UVEX9568.9/47 UVEX9568.7/45 UVEX9568.9/46 UVEX9568.9/45 UVEX9568.7/42 UVEX9568.9/44 UVEX9568.7/43 UVEX9568.7/40 UVEX9568.9/43 UVEX9568.9/42 UVEX9568.7/41 UVEX9568.8/52
ESD safety shoes, lace-up, uvex 2 xenova®, 9568
Sko og støvler
Metal-free, lightweight, flexible and fashionable safety shoe made from synthetic materials, soft padded collar and slip-resistant Xenova® dual-density PU sole resistant to heat, chemicals, oils and gasoline. All sole materials are free of silicones, plasticisers and other paint-wetting impairment substances and suitable for people allergic to chrome.
  • Compact design, anatomical shape, good lateral stability, no thermal conductivity, non-magnetic
  • Anti-twist rear cap provides more stability and kink protection
  • Climazone technology for improved comfort
  • Anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • DUO-PUR sole, non-marking, with excellent cushioning and abrasion-resistant outsole
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Shock absorbing heel and forefoot
  • Fulfils the ESD requirement: Discharge resistance <35 megohm
Ideal for use in areas where exposure to moisture is expected.
Colour: Black/green
Sertifiseringer: EN 61340 (ESD); EN ISO 20345:2011; DGUV 112-191; S2, SRC