Capsule filters, Polycap™ HD, Whatman™
Supplier: Whatman products (Cytiva)
Certificates
About this item
These Polycap™ HD capsule filters offer a broad range of particle size retention and excellent filtrate purity using polypropylene (PP) depth media. The all PP construction also offers chemical resistance for solvent filtration.
- PP filters support use with a broad range of solutions, pH solutions, and temperatures
- Sturdy construction leads to high flow and high retention capacity
- Can be autoclaved with steam at 121 °C for 20 min
- Manual vent with Luer lock to bleed air from upstream or serve as an injection or sample port
Chemical processing applications often require a capsule filter to ensure a finishing step yielding high purity final product. Different solution temperatures and pH values can complicate filtration unless an adequate filter capsule is selected.
Due to its heat sealed construction and PP filter, Polycap™ HD (Heavy Duty) is suitable for filtering chemical solutions across wide temperature and pH ranges.
Specifications
- Catalog No:
- 515-0097
- 516-0914
- 515-0100
- 516-0972
- 515-0101
- 515-0098
- 515-0099
- 515-0104
- 515-0102
- 516-0979
- 515-0103
- 516-0980
- 516-0981
- 516-0967
- 516-0958
- 516-0968
- 516-0969
- 516-0959
- 516-0982
- 516-0970
- 516-0978
- 516-0960
- 516-0912
- Type:
- Polycap™ HD 36, PP
- Polycap™ HD 75, PP
- Polycap™ HD 36, PP
- Polycap™ HD 75, PP
- Polycap™ HD 75, PP
- Polycap™ HD 36, PP
- Polycap™ HD 36, PP
- Polycap™ HD 75, PP
- Polycap™ HD 75, PP
- Polycap™ HD 150, PP
- Polycap™ HD 75, PP
- Polycap™ HD 150, PP
- Polycap™ HD 150, PP
- Polycap™ HD 75, PP
- Polycap™ HD 36, PP
- Polycap™ HD 75, PP
- Polycap™ HD 75, PP
- Polycap™ HD 36, PP
- Polycap™ HD 150, PP
- Polycap™ HD 75, PP
- Polycap™ HD 150, PP
- Polycap™ HD 36, PP
- Polycap™ HD 36, PP
- Filtration area:
- 400 cm²
- 820 cm²
- 400 cm²
- 820 cm²
- 820 cm²
- 400 cm²
- 400 cm²
- 820 cm²
- 820 cm²
- 1650 cm²
- 820 cm²
- 1650 cm²
- 1650 cm²
- 820 cm²
- 400 cm²
- 820 cm²
- 820 cm²
- 400 cm²
- 1650 cm²
- 820 cm²
- 1650 cm²
- 400 cm²
- 400 cm²
- Max. pressure:
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
Specifications
Product Information
- Catalog #Supplier #TypePore sizeFiltration areaConnectionPkAvailabilityPrice
- Specifications:
More
Specifications
TypePolycap™ HD 36, PPEnvironmentally PreferableFiltration area400 cm²Max. pressure4,1 bar - Specifications:
More
Specifications
TypePolycap™ HD 36, PPEnvironmentally PreferableFiltration area400 cm²Max. pressure4,1 bar - 515-00976703-3610Polycap™ HD 36, PP1,0 µm400 cm²Stepped barb for 6 - 10 mm tubing1Log in for availability1 * 1 Itemskr 1 575,00Specifications:
More
Specifications
TypePolycap™ HD 36, PPEnvironmentally PreferableFiltration area400 cm²Max. pressure4,1 bar - Specifications:
More
Specifications
TypePolycap™ HD 36, PPEnvironmentally PreferableFiltration area400 cm²Max. pressure4,1 bar - 515-01006703-3650Polycap™ HD 36, PP5,0 µm400 cm²Stepped barb for 6 - 10 mm tubing1Log in for availability1 * 1 Itemskr 1 575,00Specifications:
More
Specifications
TypePolycap™ HD 36, PPEnvironmentally PreferableFiltration area400 cm²Max. pressure4,1 bar - 515-00986703-3611Polycap™ HD 36, PP10 µm400 cm²Stepped barb for 6 - 10 mm tubing1Log in for availability1 * 1 Itemskr 1 575,00Specifications:
More
Specifications
TypePolycap™ HD 36, PPEnvironmentally PreferableFiltration area400 cm²Max. pressure4,1 bar - Specifications:
More
Specifications
TypePolycap™ HD 36, PPEnvironmentally PreferableFiltration area400 cm²Max. pressure4,1 bar - 515-00996703-3621Polycap™ HD 36, PP20 µm400 cm²Stepped barb for 6 - 10 mm tubing1Log in for availability1 * 1 Itemskr 1 575,00Specifications:
More
Specifications
TypePolycap™ HD 36, PPEnvironmentally PreferableFiltration area400 cm²Max. pressure4,1 bar - 516-09672710Polycap™ HD 75, PP0,45 µm820 cm²Hose barb5Log in for availability1 * 5 Itemskr 10 150,00Specifications:
More
Specifications
TypePolycap™ HD 75, PPEnvironmentally PreferableFiltration area820 cm²Max. pressure4,1 bar - Specifications:
More
Specifications
TypePolycap™ HD 75, PPEnvironmentally PreferableFiltration area820 cm²Max. pressure4,1 bar - 515-01016703-7510Polycap™ HD 75, PP1,0 µm820 cm²Stepped barb for 10 - 12 mm tubing1Log in for availability1 * 1 Itemskr 2 250,00Specifications:
More
Specifications
TypePolycap™ HD 75, PPEnvironmentally PreferableFiltration area820 cm²Max. pressure4,1 bar - Specifications:
More
Specifications
TypePolycap™ HD 75, PPEnvironmentally PreferableFiltration area820 cm²Max. pressure4,1 bar - Specifications:
More
Specifications
TypePolycap™ HD 75, PPEnvironmentally PreferableFiltration area820 cm²Max. pressure4,1 bar - 515-01046703-7550Polycap™ HD 75, PP5,0 µm820 cm²Stepped barb for 10 - 12 mm tubing1Log in for availability1 * 1 Itemskr 2 250,00Specifications:
More
Specifications
TypePolycap™ HD 75, PPEnvironmentally PreferableFiltration area820 cm²Max. pressure4,1 bar - Specifications:
More
Specifications
TypePolycap™ HD 75, PPEnvironmentally PreferableFiltration area820 cm²Max. pressure4,1 bar - 515-01026703-7511Polycap™ HD 75, PP10 µm820 cm²Stepped barb for 10 - 12 mm tubing1Log in for availability1 * 1 Itemskr 2 250,00Specifications:
More
Specifications
TypePolycap™ HD 75, PPEnvironmentally PreferableFiltration area820 cm²Max. pressure4,1 bar - Specifications:
More
Specifications
TypePolycap™ HD 75, PPEnvironmentally PreferableFiltration area820 cm²Max. pressure4,1 bar - 515-01036703-7521Polycap™ HD 75, PP20 µm820 cm²Stepped barb for 10 - 12 mm tubing1Log in for availability1 * 1 Itemskr 2 250,00Specifications:
More
Specifications
TypePolycap™ HD 75, PPEnvironmentally PreferableFiltration area820 cm²Max. pressure4,1 bar - 516-09782810TPolycap™ HD 150, PP0,45 µm1650 cm²FNPT5Log in for availability1 * 5 Itemskr 12 700,00Specifications:
More
Specifications
TypePolycap™ HD 150, PPEnvironmentally PreferableFiltration area1650 cm²Max. pressure4,1 bar - 516-09792812TPolycap™ HD 150, PP5,0 µm1650 cm²FNPT5Log in for availability1 * 5 Itemskr 12 700,00Specifications:
More
Specifications
TypePolycap™ HD 150, PPEnvironmentally PreferableFiltration area1650 cm²Max. pressure4,1 bar - 516-09812813TPolycap™ HD 150, PP10 µm1650 cm²FNPT5Log in for availability1 * 5 Itemskr 12 700,00Specifications:
More
Specifications
TypePolycap™ HD 150, PPEnvironmentally PreferableFiltration area1650 cm²Max. pressure4,1 bar - 516-09802813Polycap™ HD 150, PP10 µm1650 cm²Hose barb5Log in for availability1 * 5 Itemskr 12 500,00Specifications:
More
Specifications
TypePolycap™ HD 150, PPEnvironmentally PreferableFiltration area1650 cm²Max. pressure4,1 bar - 516-09822814TPolycap™ HD 150, PP20 µm1650 cm²FNPT5Log in for availability1 * 5 Itemskr 12 700,00Specifications:
More
Specifications
TypePolycap™ HD 150, PPEnvironmentally PreferableFiltration area1650 cm²Max. pressure4,1 bar