Pesticide reference standard, Imazapic, PESTANAL®, analytical standard, Supelco®
Leverandør: Merck
PESTANAL®
34179-100MGEA 1225 NOK
34179-100MG
Pesticide reference standard, Imazapic, PESTANAL®, analytical standard, Supelco®
Standarder Organiske standarder
Utilised as a reference standard for the determination of imazapic in rice samples by high-performance liquid chromatography.
  • Imazapic is an imidazolinone herbicide
  • Effective against weedy rice constraints